Få mere at vide
1H-Pyrazole-3,4,5-d3, TRC
Artikelnummer. 30427031
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30427031 250 mg
30421898 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427031 Leverandør Toronto Research Chemicals Leverandørnr. TRCP842197250MG
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 51038-79-0
Molekylær formel C3HD3N2
Molekylvægt (g/mol) 71.1
Synonym 3,4,5-Trideuteropyrazole, Pyrazole-3,4,5-d3
SMIL [2H]C1=C([2H])C([2H])=NN1
Tekniske data
Kemisk navn eller materiale 1H-Pyrazole-3,4,5-d3
Mængde 250 mg
InChI formel InChI=1S/C3H4N2/c1-2-4-5-3-1/h1-3H,(H,4,5)/i1D,2D,3D
Formel vægt 71.0563