missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole, 99%, Thermo Scientific Chemicals

Catalog Number p-63569
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
95-14-7
C6H5N3
119.13
MFCD00005699
QRUDEWIWKLJBPS-UHFFFAOYSA-N
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
7220
CHEBI:75331
2H-benzotriazole
C1=CC2=NNN=C2C=C1
This item is not returnable. View return policy
95-14-7
C6H5N3
119.13
MFCD00005699
QRUDEWIWKLJBPS-UHFFFAOYSA-N
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
7220
CHEBI:75331
2H-benzotriazole
C1=CC2=NNN=C2C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 94.0°C to 99.0°C
Color Beige-Yellow to White
Boiling Point 204.0°C (15.0 mmHg)
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 26, 38
Fieser 16,21
Merck Index 15, 1111
Solubility Information Solubility in water: 25g/L in water (20°C). Other solubilities: soluble in alcohol,benzene,toluene,chloroform,and dmf,soluble in ethanol,methanol and ethyleneglycol
Formula Weight 119.13
Percent Purity 99%
Physical Form Powder, Granules, Crystals, Needles or Flakes
Chemical Name or Material 1H-Benzotriazole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1H-Benzotriazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'