Få mere at vide
1H-Benzimidazole-2-sulfonic Acid, Mikromol™
Artikelnummer. 30051969
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051969 100 mg
30049975 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051969 Leverandør Mikromol Leverandørnr. MM0618.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1H-Benzimidazole-2-sulfonic Acid
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1H-Benzimidazole-2-sulfonic Acid
CAS 40828-54-4
Analyt- eller komponentnavne 1H-Benzimidazole-2-sulfonic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C7 H6 N2 O3 S
Certificeringer/compliance ISO 17025
Synonym 1H-Benzimidazole-2-sulfonic acid, 2-Benzimidazolesulfonic acid
InChI formel InChI=1S/C7H6N2O3S/c10-13(11,12)7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H,8,9)(H,10,11,12)
SMIL OS(=O)(=O)c1nc2ccccc2[nH]1
IUPAC navn 1H-benzimidazole-2-sulfonic acid
Molekylvægt (g/mol) 198.2
Mængde 100 mg
Formel vægt 198.0099 g/mol