Få mere at vide
1H,1H-Perfluorooctyl acrylate, 97%, stab. with 50ppm 4-methoxyphenol, Thermo Scientific™
Artikelnummer. 11369275
Skift visning
Mængde:
1 g
5 g
25 g
Pakningsstørrelse:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11369275 5 g 5g
11359275 1 g 1g
11379275 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11369275 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L16603.06
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 307-98-2
Molekylær formel C11H5F15O2
Molekylvægt (g/mol) 454.135
MDL nummer MFCD00039248
InChI nøgle YSQGYEYXKXGAQA-UHFFFAOYSA-N
Synonym 1h,1h-pentadecafluorooctyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl acrylate, 1h,1h-perfluorooctyl acrylate, acrylic acid 1h,1h-pentadecafluoro-n-octyl ester, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl acrylate, 1h,1h-pentadecafluoro-n-octyl acrylate, 1,1-dihydroperfluorooctyl acrylate, 1,1-dyhydroperfluorooctyl acrylate, acrylic acid 1h,1h-perfluorooctyl ester
PubChem CID 67551
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
SMIL C=CC(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Tekniske data
Kemisk navn eller materiale 1H,1H-Perfluorooctyl acrylate
Navn note stabilized with 50ppm 4-methoxyphenol
Tæthed 1.403
Kogepunkt 112°C to 115°C (60 mmHg)
Flammepunkt 91°C (195°F)
Lineær formel CF3(CF2)6CH2O2CCH=CH2
Brydningsindeks 1.3275
Mængde 5 g
Beilstein 1809801
Formel vægt 454.14
Procent renhed 97%