missing translation for 'onlineSavingsMsg'
Få mere at vide

1H-1,2,4-Triazole-1-acetic acid, 97%

Artikelnummer. p-7030855
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15464357 250 mg 250mg
15454357 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15464357 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H31732.MD
Request Bulk or Custom Format

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

CAS: 28711-29-7 | C4H5N3O2 | 127.103 g/mol

1H-1,2,4-Triazole-1-acetic acid is used to produce (1-hydroxy-1-phosphono-2-[1,2,4]triazol-1-yl-ethyl)-phosphonic acid at the temperature of 100 - 110°C. It will need reagent H3PO4, PCl3 and solvent chlorobenzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-1,2,4-Triazole-1-acetic acid is used to produce (1-hydroxy-1-phosphono-2-[1,2,4]triazol-1-yl-ethyl)-phosphonic acid at the temperature of 100 - 110°C. It will need reagent H3PO4, PCl3 and solvent chlorobenzene

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 28711-29-7
Molekylær formel C4H5N3O2
Molekylvægt (g/mol) 127.103
MDL nummer MFCD00036051
InChI nøgle RXDBSQXFIWBJSR-UHFFFAOYSA-N
Synonym 1,2,4-triazole-1-acetic acid, 2-1h-1,2,4-triazol-1-yl acetic acid, 1h-1,2,4-triazole-1-acetic acid, 1,2,4 triazol-1-yl-acetic acid, unii-w3ask2wbnh, 1,2,4-triazol-1-ylacetic acid, w3ask2wbnh, 1h-1,2,4-triazol-1-ylacetic acid, 1h-1,2,4-triazol-1-yl acetic acid, 1h-1,2,4-triazole-1-aceticacid
PubChem CID 1810180
IUPAC navn 2-(1,2,4-triazol-1-yl)acetic acid
SMIL C1=NN(C=N1)CC(=O)O

Tekniske data

Kemisk navn eller materiale 1H-1,2,4-Triazole-1-acetic acid
Smeltepunkt 201°C to 205°C
Mængde 250 mg
Opløselighedsinformation Soluble in water.
Formel vægt 127.1
Procent renhed 97%

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.