Få mere at vide
1H-1,2,4-triazol-5-ol, TRC
Artikelnummer. 30407681
Skift visning
Mængde:
500 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30407681 500 mg -
30390373 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30407681 Leverandør Toronto Research Chemicals Leverandørnr. TRCH953870500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1H-1,2,4-triazol-5-ol
CAS 930-33-6
Molekylær formel C2H3N3O
Mængde 500 mg
Synonym 1,2-Dihydro-3H-1,2,4-triazol-3-one (ACI), s-Triazol-3-ol (7CI), Δ2-1,2,4-Triazolin-5-one (6CI, 8CI), 1,2-Dihydro-1,2,4-triazol-3-one, 1,2-Dihydro-3H-1,2,4-triazole-3-one, 1H-1,2,4-Triazol-3-ol, 2,4-Dihydro-3H-1,2,4-triazol-3-one, NSC 119861, NSC 153380, NOA 457654, 1H-1,2,4-Triazol-5-ol
InChI formel InChI=1S/C2H3N3O/c6-2-3-1-4-5-2/h1H,(H2,3,4,5,6)
SMIL Oc1nc[nH]n1
IUPAC navn 1H-1,2,4-triazol-3-ol
Molekylvægt (g/mol) 85.06
Formel vægt 85.0276