Få mere at vide
(1E)-Tributyl Aconitate, TRC
Artikelnummer. 30451239
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30451239 500 mg
1 options
Artikelnummer. 30451239 Leverandør Toronto Research Chemicals Leverandørnr. TRCT756895500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tributyl (E)-Propene-1,2,3-tricarboxylate (Tributyl (E)-Aconitate)
CAS 343599-72-4
Molekylær formel C18 H30 O6
Mængde 500 mg
Synonym Tributyl Acetylcitrate Imp. B (EP) (E-Isomer), Tributyl (E)-Propene-1,2,3-tricarboxylate, Tributyl (E)-Aconitate, Tributyl Acetylcitrate Impurity B as (E)-Isomer
InChI formel InChI=1S/C18H30O6/c1-4-7-10-22-16(19)13-15(18(21)24-12-9-6-3)14-17(20)23-11-8-5-2/h13H,4-12,14H2,1-3H3/b15-13+
SMIL O=C(/C=C(C(OCCCC)=O)\CC(OCCCC)=O)OCCCC
IUPAC navn tributyl (E)-prop-1-ene-1,2,3-tricarboxylate
Molekylvægt (g/mol) 342.43
Formel vægt 342.2042