Få mere at vide
1Alpha-Hydroxy Vitamin D2-d3, TRC
Artikelnummer. 30407773
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30407773 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407773 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9958021MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1alpha-Hydroxy Vitamin D2-d3
CAS 163081-47-8
Molekylær formel C28H41D3O2
Mængde 1 mg
Synonym (1α,3β,5Z,7E,22E)- 9,10-Secoergosta-5,7,10(19),22-tetraene-1,3-diol-d3
InChI formel InChI=1S/C28H44O2/c1-18(2)19(3)9-10-20(4)25-13-14-26-22(8-7-15-28(25,26)6)11-12-23-16-24(29)17-27(30)21(23)5/h9-12,18-20,24-27,29-30H,5,7-8,13-17H2,1-4,6H3/b10-9+,22-11+,23-12-/t19?,20-,24-,25-,26+,27+,28-/m1/s1/i3D3
SMIL [H][C@]1([C@H](C)/C=C/C(C([2H])([2H])[2H])C(C)C)CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)C[C@@H]3O)/C3=C
IUPAC navn (1R,3S,Z)-5-(2-((1R,3aS,7aR,E)-7a-methyl-1-((2R,E)-6-methyl-5-(methyl-d3)hept-3-en-2-yl)octahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol
Molekylvægt (g/mol) 415.67
Formel vægt 415.35