Få mere at vide
(19E/Z)-seco Rapamycin Sodium Salt (>80%), TRC
Artikelnummer. 30138338
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138338 100 mg
30136073 50 mg
30011301 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138338 Leverandør Toronto Research Chemicals Leverandørnr. TRCR124010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148554-65-8
Molekylær formel C51H78NNaO13
Molekylvægt (g/mol) 936.15
Synonym (2E,4R,6E,8R)-1-[2-Oxo-2-[(2R,3R,6S)-tetrahydro-2-hydroxy-6-[(2S,3E,5E,7E,9S,11R,13R,14R,15E,17R,19EZ,21R)-14-hydroxy-22-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-2,13-dimethoxy-3,9,11,15,17,21-hexamethyl-12,18-dioxo-3,5,7,15,19-docosapentaen-1-yl]-3-met
IUPAC navn sodium (S)-1-(2-((2R,3R,6S)-2-hydroxy-6-((2S,3E,5E,7E,9S,11R,13R,14R,15E,17R,19E,21R)-14-hydroxy-22-((1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl)-2,13-dimethoxy-3,9,11,15,17,21-hexamethyl-12,18-dioxodocosa-3,5,7,15,19-pentaen-1-yl)-3-methyltetrahydro-2H-pyran-2-yl)-2-oxoacetyl)piperidine-2-carboxylate
SMIL C[C@H](/C=C/C=C/C=C(\C)/[C@H](C[C@@H]1CC[C@@H](C)[C@@](C(=O)C(=O)N2CCCC[C@H]2C(=O)[O-])(O)O1)OC)C[C@@H](C)C(=O)[C@@H]([C@@H](/C(=C/[C@@H](C)C(=O)/C=C/[C@H](C)C[C@@H]3CC[C@H]([C@@H](C3)OC)O)/C)O)OC.[Na+]
Tekniske data
Kemisk navn eller materiale (19EZ)-seco-Rapamycin Sodium
Mængde 100 mg
InChI formel InChI=1S/C51H79NO13.Na/c1-31(26-35(5)45(55)47(64-10)46(56)36(6)28-34(4)41(53)23-19-32(2)27-38-21-24-42(54)44(29-38)63-9)16-12-11-13-17-33(3)43(62-8)30-39-22-20-37(7)51(61,65-39)48(57)49(58)52-25-15-14-18-40(52)50(59)60;/h11-13,16-17,19,23,28,31-32,34-35,37-40,42-44,46-47,54,56,61H,14-15,18,20-22,24-27,29-30H2,1-10H3,(H,59,60);/q;+1/p-1/b13-11+,16-12+,23-19+,33-17+,36-28+;/t31-,32+,34-,35-,37-,38+,39+,40+,42-,43+,44-,46-,47+,51-;/m1./s1
Formel vægt 935.54