Få mere at vide
(±)-18-Methyl-d3-eicosanoic Acid, CDN
Artikelnummer. 30316998
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316998 0.05 g -
30327947 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316998 Leverandør CDN Leverandørnr. CDND75330.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 18-Methyleicosanoic Acid-D3 (18-methyl-D3)
CAS 2708278-14-0
Molekylær formel C21 D3 H39 O2
Mængde 0.05 g
Synonym (±)-18-Methyl-d3-eicosanoic Acid, 18-(Methyl-d3)eicosanoic acid, 18-Methylicosanoic acid-d3, 18-MEA-d3, Trideuteromethylarachidic Acid, 18-Methyleicosanoic Acid-D3 (18-methyl-D3)
InChI formel InChI=1 S/C21H42O2/c1-3-20(2)18-16-14-12-10-8-6-4-5-7-9-11-13-15-17-19-21(22)23/h20H,3-19H2,1-2H3,(H,22,23)/i2D3
SMIL [2 H]C([2 H])([2 H])C(CC)CCCCCCCCCCCCCCCCC(=O)O
IUPAC navn 18-(trideuteriomethyl)icosanoic acid
Molekylvægt (g/mol) 329.5755
Formel vægt 329.3373 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity