missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(17R)-4'-Chloro-5'-ethyl-9-fluoro-11beta-hydroxy-16beta-methylspiro[androsta-1,4-diene-17,2'(3'H)-furan]-3,3'-dione, Mikromol
Description
(17R)-4'-Chloro-5'-ethyl-9-fluoro-11beta-hydroxy-16beta-methylspiro[androsta-1,4-diene-17,2'(3'H)-furan]-3,3'-dione, Mikromol
Specifications
Specifications
| Kemisk navn eller materiale | Clobetasol Propionate 17alpha-spiro Compound |
| CAS | 1486466-31-2 |
| Analyt- eller komponentnavne | Clobetasol Propionate 17alpha-spiro Compound |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H30 Cl F O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | (17R)-4′-Chloro-5′-ethyl-9-fluoro-11β-hydroxy-16β-methylspiro[androsta-1,4-diene-17,2′(3′H)-furan]-3,3′-dione, Clobetasol Propionate 17α-spiro Compound, (17R)-4'-Chloro-5'-ethyl-9-fluoro-11beta-hydroxy-16beta-methylspiro[androsta-1,4-diene-17,2'(3'H)-furan]-3,3'-dione, Clobetasol Propionate 17alpha-spiro Compound, 9a-Fluoro-11b-hydroxy-16b-methyl-3-oxoandrosta-1,4-diene-17(R)-spiro-2'-[4'-chloro-5'-ethylfuran-3'(2'H)-one, USP Clobetasol propionate Related Compound A, Ph Eur Clobetasol Propionate Impurity J, CCI18747, CCI6990, Clobetasol-17-propionate, CCI1822, Clobetasol-17-propionate isomer, 21-desoxybetamethasone-17-propionate, 16-ene, Ethylenic acid, USP Clobetasol propionate Related Compound A |
| InChI formel | InChI=1S/C25H30ClFO4/c1-5-18-20(26)21(30)25(31-18)13(2)10-17-16-7-6-14-11-15(28)8-9-22(14,3)24(16,27)19(29)12-23(17,25)4/h8-9,11,13,16-17,19,29H,5-7,10,12H2,1-4H3/t13-,16-,17-,19-,22-,23-,24-,25-/m0/s1 |
| Løsningstype | Neat |
| SMIL | CCC1=C(Cl)C(=O)[C@@]2(O1)[C@@H](C)C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?