Få mere at vide
(17R)-4’-Chloro-5’-ethyl-9-fluoro-11Beta-hydroxy-16Beta-methylspiro[androsta-1,4-diene-17,2’(3’H)-fu, TRC
Artikelnummer. 30006342
Skift visning
Mængde:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006342 250 mg
18053030 25 mg
18061120 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006342 Leverandør Toronto Research Chemicals Leverandørnr. TRCC366115250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1486466-31-2
Molekylær formel C25 H30 Cl F O4
Molekylvægt (g/mol) 448.95
Synonym (17R)-4'-Chloro-5'-ethyl-9-fluoro-11β-hydroxy-16β-methylspiro[androsta-1,4-diene-17,2'(3'H)-furan]-3,3'-dione, Clobetasol Propionate 17α-spiro Compound, 9α-Fluoro-11β-hydroxy-16β-methyl-3-oxoandrosta-1,4-diene-17(R)-spiro-2'-[4'-chloro-5'-ethylfuran-3'(2'H)-one
IUPAC navn (8S,9R,10S,11S,13S,14S,16S,17R)-4'-chloro-5'-ethyl-9-fluoro-11-hydroxy-10,13,16-trimethylspiro[6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17,2'-furan]-3,3'-dione
SMIL CCC1=C(Cl)C(=O)[C@@]2(O1)[C@@H](C)C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C
Tekniske data
Kemisk navn eller materiale Clobetasol Propionate 17alpha-spiro Compound
Analyt- eller komponentnavne Clobetasol Propionate 17alpha-spiro Compound
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C25H30ClFO4/c1-5-18-20(26)21(30)25(31-18)13(2)10-17-16-7-6-14-11-15(28)8-9-22(14,3)24(16,27)19(29)12-23(17,25)4/h8-9,11,13,16-17,19,29H,5-7,10,12H2,1-4H3/t13-,16-,17-,19-,22-,23-,24-,25-/m0/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 250 mg
Formel vægt 448.1817