Få mere at vide
17beta-Dihydroequilin-16,16,17-d3, CDN
Artikelnummer. 30323858
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323858 0.005 g
30315095 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323858 Leverandør CDN Leverandørnr. CDND42600.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 17 beta-Dihydroequilin-16,16,17-d3
CAS 350820-03-0
Molekylær formel C18 2H3 H17 O2
Mængde 0.005 g
InChI formel InChI=1 S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16-17,19-20 H,6-9H2,1H3/t16-,17-,18-/m0/s1/i7D2,17 D
SMIL [2 H]C1([2 H])C[C@H]2c3ccc4cc(O)ccc4c3CC[C@]2(C)[C@@]1([2 H])O
IUPAC navn (13 S,14 S,17 S)-16,16,17-trideuterio-13-methyl-11,12,14,15-tetrahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 271.37
Formel vægt 271.1652 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity