missing translation for 'onlineSavingsMsg'
Få mere at vide

17beta-Dihydro Equilin 3-Sulfate Sodium Salt, TRC

Artikelnummer. 30378030
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30378030 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30378030 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44897025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 17beta-Dihydro Equilin 3-Sulfate Sodium Salt
CAS 16680-49-2
Molekylær formel C18 H21 O5 S . Na
Mængde 25 mg
Synonym Estra-1,3,5(10),7-tetraene-3,17-diol, 3-(hydrogen sulfate), sodium salt (1:1), Estra-1,3,5(10),7-tetraene-3,17-diol, 3-(hydrogen sulfate), monosodium salt, (17β)- (9CI), Estra-1,3,5(10),7-tetraene-3,17β-diol, 3-(hydrogen sulfate), monosodium salt (8CI), 17β-Dihydroequilin sodium sulfate, Sodium 17β-dihydroequilin sulfate
InChI formel InChI=1S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3-5,10,14,16-17,19H,2,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,16+,17+,18+;/m1./s1
SMIL [Na+].C[C@]12CC[C@H]3C(=CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CC[C@@H]2O
IUPAC navn sodium;[(9S,13S,14S,17S)-17-hydroxy-13-methyl-6,9,11,12,14,15,16,17-octahydrocyclopenta[a]phenanthren-3-yl] sulfate
Molekylvægt (g/mol) 372.41
Formel vægt 372.1007
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.