Få mere at vide
17Beta-Dihydro Equilin-16,16,17-d3, TRC
Artikelnummer. 30378031
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30378031 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378031 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4491321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 17beta-Dihydroequilin-16,16,17-d3
CAS 350820-03-0
Molekylær formel C18 2H3 H17 O2
Mængde 1 mg
Synonym Estra-1,3,5(10),7-tetraene-16,16,17-d3-3,17-diol, (17β)- (9CI), (17β)-Estra-1,3,5(10),7-tetraene-3,17-diol-16,16,17-d3, 17β-Dihydroequilin-d3, 17β-Dihydroequilin-16,16,17-D3, 7-Dehydroestradiol-16,16,17-d3, β-Dihydroequilin-d3
InChI formel InChI=1S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h2-5,10,16-17,19-20H,6-9H2,1H3/t16-,17-,18-/m0/s1/i7D2,17D
SMIL [2H]C1([2H])C[C@H]2c3ccc4cc(O)ccc4c3CC[C@]2(C)[C@@]1([2H])O
IUPAC navn (13S,14S,17S)-16,16,17-trideuterio-13-methyl-11,12,14,15-tetrahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 271.37
Formel vægt 271.1652