Få mere at vide
17Beta-Dihydro-17Alpha-ethynyl-equilenin, TRC
Artikelnummer. 30134168
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134168 10 mg
17912845 1 mg
17925035 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134168 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44759010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1357266-17-1
Molekylær formel C20H20O2
Molekylvægt (g/mol) 292.37
Synonym (17alpha)-19-Norpregna-1,3,5,7,9-pentaen-20-yne-3,17-diol
SMIL C[C@]12CCc3c(ccc4cc(O)ccc34)[C@@H]1CC[C@@]2(O)C#C
Tekniske data
Kemisk navn eller materiale 17beta-Dihydro-17alpha-ethynyl-equilenin
Mængde 10 mg
InChI formel InChI=1S/C20H20O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,4-7,12,18,21-22H,8-11H2,2H3/t18-,19-,20-/m0/s1
Formel vægt 292.1463