Få mere at vide
17Alpha-Progesterone, TRC
Artikelnummer. 30006012
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006012 1 mg
30007436 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006012 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7558951MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2000-66-0
Molekylær formel C21 H30 O2
Molekylvægt (g/mol) 314.46
Synonym (8S,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-1,2,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-3H-cyclopenta[a]phenanthren-3-one, Pregn-4-ene-3,20-dione, (17α)-, 17α-Pregn-4-ene-3,20-dione (6CI,7CI,8CI), (17α)-Pregn-4-ene-3,20-dione, 17-Isoprogesterone, 17α-Progesterone, Isoprogesterone, Progesterone Imp. M (EP);Progesterone EP Impurity M
IUPAC navn (8S,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one
SMIL CC(=O)[C@@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 17alpha-Progesterone
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h12,16-19H,4-11H2,1-3H3/t16-,17-,18-,19-,20-,21+/m0/s1
Formel vægt 314.2246
Procent renhed >95%