Få mere at vide
17Alpha-Hydroxy Progesterone, TRC
Artikelnummer. 17902615
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17902615 5 g
17982905 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17902615 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9523305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68-96-2
Molekylær formel C21 H30 O3
Molekylvægt (g/mol) 330.46
Synonym Pregn-4-ene-3,20-dione, 17-hydroxy-, 17-Hydroxypregn-4-ene-3,20-dione, 17-Hydroxyprogesterone, 17α-Hydroxy-P4, 17α-Hydroxylutin, 17α-Hydroxypregn-4-ene-3,20-dione, 17α-Hydroxyprogesterone, Gestageno, Gestageno Gador, Hydroxyprogesterone, NSC 15468, Pregn-4-en-17α-ol-3,20-dione, Prodix, Prodox, U 3096, δ4-Pregnen-17α-ol-3,20-dione
IUPAC navn (8R,9S,10R,13S,14S,17R)-17-acetyl-17-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
SMIL CC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 17a-Hydroxyprogesterone
Mængde 5 g
InChI formel InChI=1S/C21H30O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h12,16-18,24H,4-11H2,1-3H3/t16-,17+,18+,19+,20+,21+/m1/s1
Formel vægt 330.2195