Få mere at vide
17alpha-Ethynylestradiol-2,4,16,16-d4, CDN
Artikelnummer. 30321510
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321510 0.01 g
30313166 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321510 Leverandør CDN Leverandørnr. CDND43190.01G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethynyl Estradiol-2,4,16,16-d4
CAS 350820-06-3
Molekylær formel C20 2H4 H20 O2
Mængde 0.01 g
Synonym 19-Norpregna-1,3,5(10)-trien-20-yne-2,4,16,16-d4-3,17-diol, (17α)-, (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-2,4,16,16-d4-3,17-diol, Ethynyl estradiol-2,4,16,16-d4, Ethinylestradiol (2,4,16,16-D4)
InChI formel InChI=1 S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22 H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1/i5D,11D2,12 D
SMIL [2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@@]4(O)C#C)[C@@H]3 CCc2c([2 H])c1O
IUPAC navn (8 R,9 S,13 S,14 S,17 R)-2,4,16,16-tetradeuterio-17-ethynyl-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 300.43
Formel vægt 300.2027 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity