Få mere at vide
17Alpha-Des-(2-hydroxyacetyl)budesonide 17Alpha-Carboxylic Acid, TRC
Artikelnummer. 30138738
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138738 10 mg
17902005 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138738 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28989510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 192057-49-1
Molekylær formel C24H32O6
Molekylvægt (g/mol) 416.51
Synonym Androsta-1,4-diene-17-carboxylic acid, 16,17-[butylidenebis(oxy)]-11-hydroxy-3-oxo-, (11β,16α,17α)-, (11β,16α,17α)-16,17-[Butylidenebis(oxy)]-11-hydroxy-3-oxoandrosta-1,4-diene-17-carboxylic acid, Budesonide 20-Carboxylic Acid
IUPAC navn (1S,2S,4R,8S,9S,11S,12S,13R)-11-hydroxy-9,13-dimethyl-16-oxo-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-diene-8-carboxylic acid
SMIL CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)O
Tekniske data
Kemisk navn eller materiale 17a-Des-(2-hydroxyacetyl)budesonide 17a-Carboxylic Acid
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C24H32O6/c1-4-5-19-29-18-11-16-15-7-6-13-10-14(25)8-9-22(13,2)20(15)17(26)12-23(16,3)24(18,30-19)21(27)28/h8-10,15-20,26H,4-7,11-12H2,1-3H3,(H,27,28)/t15-,16-,17-,18+,19?,20+,22-,23-,24-/m0/s1
Formel vægt 416.2199
Procent renhed >95%