Få mere at vide
17a-Ethinylestradiol D4 (2,4,16,16-D4) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Artikelnummer. 30314202
Skift visning
Mængde:
1 mL
Artikelnummer. Mængde
30314202 1 mL
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314202 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA13245105AL100
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Ethynyl Estradiol-2,4,16,16-d4
CAS 350820-06-3
Opløsningsmiddel eller Matrix Acetonitrile
Molekylær formel C20 2H4 H20 O2
Mængde 1 mL
Synonym 19-Norpregna-1,3,5(10)-trien-20-yne-2,4,16,16-d4-3,17-diol, (17α)-, (17α)-19-Norpregna-1,3,5(10)-trien-20-yne-2,4,16,16-d4-3,17-diol, Ethynyl estradiol-2,4,16,16-d4, Ethinylestradiol (2,4,16,16-D4)
InChI formel InChI=1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20+/m1/s1/i5D,11D2,12D
SMIL [2H]c1cc2[C@H]3CC[C@@]4(C)[C@@H](CC([2H])([2H])[C@@]4(O)C#C)[C@@H]3CCc2c([2H])c1O
IUPAC navn (8R,9S,13S,14S,17R)-2,4,16,16-tetradeuterio-17-ethynyl-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvægt (g/mol) 300.43
Formel vægt 300.2027 g/mol