Learn More
Hamilton™ 1700 Series Removable Needle Syringes
Ideal for dispensing both liquids and gases.
683.00 DKK - 11865.00 DKK
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Price | Quantity & Availability | |||||
11507804
|
Hamilton™
80014 |
32 G |
683.00 DKK
Each |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11528024
|
Hamilton™
DS80065 |
22s G |
11865.00 DKK
Each |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Precision-machined PTFE plunger tip which creates a leak-free seal
- Tip wipes the interior of the syringe barrel free of sample
- Fluid path is chemically inert as possible
Specifications
10 μL |