Få mere at vide
17-O-Acetyl Diflorasone, TRC
Artikelnummer. 30137420
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137420 50 mg
30134003 100 mg
30010446 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137420 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17367050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50630-19-8
Molekylær formel C24H30F2O6
Molekylvægt (g/mol) 452.49
Synonym 17-Acetoxy-6alpha,9-difluoro-11beta,21-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione;Diflorasone 17-Acetate;
IUPAC navn [(6S,8R,9S,10S,11R,13S,14S,16S,17S)-6,9-difluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] acetate
SMIL C[C@H]1C[C@H]2[C@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@]3(F)[C@H](O)C[C@]2(C)[C@]1(OC(=O)C)C(=O)CO
Tekniske data
Kemisk navn eller materiale 17-O-Acetyl Diflorasone
Mængde 50 mg
InChI formel InChI=1S/C24H30F2O6/c1-12-7-15-16-9-18(25)17-8-14(29)5-6-21(17,3)23(16,26)19(30)10-22(15,4)24(12,20(31)11-27)32-13(2)28/h5-6,8,12,15-16,18-19,27,30H,7,9-11H2,1-4H3/t12-,15-,16+,18-,19+,21-,22-,23+,24+/m0/s1
Formel vægt 452.201