Få mere at vide
17-Methyl-5Alpha-androstane-3Beta,17Beta-diol-d3, TRC
Artikelnummer. 30483122
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30483122 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30483122 Leverandør Toronto Research Chemicals Leverandørnr. TRCM28785210MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 17-Methyl-5alpha-androstane-3beta,17beta-diol-d3
CAS 88247-85-2
Molekylær formel C20H31D3O2
Mængde 10 mg
Synonym (3beta,5alpha,17beta)-17-(Methyl-d3)androstane-3,17-diol, 17alpha-(Methyl-d3)androst-4-ene-3beta,17beta-diol, 17alpha-(Methyl-d3)androstan-3beta-17beta-diol, 17beta-Hydroxy-17alpha-(methyl-d3)5alpha-androstan-3beta-ol, 5alpha-Androstane-17alpha-(methyl-d3)-3beta,17beta-diol, NSC 118175
InChI formel InChI=1S/C20H34O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h13-17,21-22H,4-12H2,1-3H3/t13?,14-,15+,16-,17-,18-,19-,20-/m0/s1/i3D3
SMIL [2H]C([2H])([2H])[C@]1(O)CC[C@H]2[C@@H]3CCC4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Molekylvægt (g/mol) 309.5