Få mere at vide
17-Desoxy-Dexamethasone 21-Acetate, TRC
Artikelnummer. 30365110
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30365110 100 mg
30352822 5 mg
30359630 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30365110 Leverandør Toronto Research Chemicals Leverandørnr. TRCD296985100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1597-78-0
Molekylær formel C24 H31 F O5
Molekylvægt (g/mol) 418.5
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-fluoro-11-hydroxy-16-methyl-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,21-dihydroxy-16α-methyl-, 21-acetate (8CI), Pregna-1,4-diene-3,20-dione, 9α-fluoro-11β,21-dihydroxy-16α-methyl-, 21-acetate (7CI), (11β,16α)-21-(Acetyloxy)-9-fluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione, 21-Acetoxy-9-fluoro-11β-hydroxy-16α-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methylpregna-1,4-diene-11β,21-diol-3,20-dione-21-acetate
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16R,17S)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@H]1C(=O)COC(=O)C
Tekniske data
Kemisk navn eller materiale Desoxymetasone 21-Acetate
Mængde 100 mg
InChI formel InChI=1S/C24H31FO5/c1-13-9-18-17-6-5-15-10-16(27)7-8-23(15,4)24(17,25)20(29)11-22(18,3)21(13)19(28)12-30-14(2)26/h7-8,10,13,17-18,20-21,29H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,20+,21-,22+,23+,24+/m1/s1
Formel vægt 418.2156