Få mere at vide
17-Deshydroxyacetyl Betamethasone 17-(Hydroxymethylcarboxylate), TRC
Artikelnummer. 30362801
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362801 50 mg
30351669 5 mg
30366325 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362801 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29828050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1184871-61-1
Molekylær formel C22H29FO6
Molekylvægt (g/mol) 408.47
Synonym (11beta,16beta)-9-Fluoro-11,17,20-trihydroxy-16-methyl-3-oxopregna-1,4-dien-21-oic Acid, Dexamethasone impurity 11, rac-2-((8S,9R,10S,11S,13S,14S,16S,17R)-9-Fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-hydroxyacetic Acid
SMIL C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(O)C(=O)O
Tekniske data
Kemisk navn eller materiale 17-Deshydroxyacetyl Betamethasone 17-(Hydroxymethylcarboxylate)
Mængde 50 mg
InChI formel InChI=1S/C22H29FO6/c1-11-8-15-14-5-4-12-9-13(24)6-7-19(12,2)21(14,23)16(25)10-20(15,3)22(11,29)17(26)18(27)28/h6-7,9,11,14-17,25-26,29H,4-5,8,10H2,1-3H3,(H,27,28)/t11-,14-,15-,16-,17?,19-,20-,21-,22-/m0/s1
Formel vægt 408.1948