Learn More
Cytiva 17 CHR Filter Paper
Brand:  Cytiva 30176715
300.20 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 0.15980kg
Product Code. 12475342
  • 316.00 DKK / Pack of 100
Estimated Shipment: 18-09-2024
to see stock.
Specifications
57 x 57 mm
17 CHR