Få mere at vide
16alpha-Hydroxy Androsterone, TRC
Artikelnummer. 30135051
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135051 100 mg
17934175 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135051 Leverandør Toronto Research Chemicals Leverandørnr. TRCH828170100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14167-49-8
Molekylær formel C19 H30 O3
Molekylvægt (g/mol) 306.44
Synonym Androstan-17-one, 3,16-dihydroxy-, (3α,5α,16α)-, 5α-Androstan-17-one, 3α,16α-dihydroxy- (8CI), (3α,5α,16α)-3,16-Dihydroxyandrostan-17-one, 16α-Hydroxyandrosterone, 3α,16α-Dihydroxy-5α-androstan-17-one, 3α,16α-Dihydroxy-5α-androstane-17-one
IUPAC navn (3R,5S,8R,9S,10S,13S,14S,16R)-3,16-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-one
SMIL C[C@]12CC[C@@H](O)C[C@@H]1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3C[C@@H](O)C4=O
Tekniske data
Kemisk navn eller materiale 16alpha-Hydroxyandrosterone
Mængde 100 mg
InChI formel InChI=1S/C19H30O3/c1-18-7-5-12(20)9-11(18)3-4-13-14(18)6-8-19(2)15(13)10-16(21)17(19)22/h11-16,20-21H,3-10H2,1-2H3/t11-,12+,13+,14-,15-,16+,18-,19-/m0/s1
Formel vægt 306.2195