Få mere at vide
16Alpha-Hydroxy-17Beta-estradiol-2,4-d2, TRC
Artikelnummer. 30373815
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30373815 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373815 Leverandør Toronto Research Chemicals Leverandørnr. TRCE8889615MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estriol-2,4-d2
CAS 53866-32-3
Molekylær formel C18 2H2 H22 O3
Mængde 5 mg
Synonym Estriol-(2,4-D2), Estra-1,3,5(10)-triene-2,4-d2-3,16,17-triol, (16α,17β)-, (16α,17β)-Estra-1,3,5(10)-triene-2,4-d2-3,16,17-triol, Estriol-2,4-d2
InChI formel InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1/i3D,8D
SMIL [2H]c1cc2[C@H]3CC[C@]4(C)[C@@H](O)[C@H](O)C[C@H]4[C@@H]3CCc2c([2H])c1O
IUPAC navn (8R,9S,13S,14S,16R,17R)-2,4-dideuterio-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
Molekylvægt (g/mol) 290.39
Formel vægt 290.1851