Få mere at vide
16alpha-Hydroxy-17beta-estradiol-2,4-d2, CDN
Artikelnummer. 30314268
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314268 0.005 g
30311185 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314268 Leverandør CDN Leverandørnr. CDND52790.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Estriol-2,4-d2
CAS 53866-32-3
Molekylær formel C18 2H2 H22 O3
Mængde 0.005 g
Synonym Estriol-(2,4-D2), Estra-1,3,5(10)-triene-2,4-d2-3,16,17-triol, (16α,17β)-, (16α,17β)-Estra-1,3,5(10)-triene-2,4-d2-3,16,17-triol, Estriol-2,4-d2
InChI formel InChI=1 S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21 H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1/i3D,8 D
SMIL [2 H]c1cc2[C@H]3 CC[C@]4(C)[C@@H](O)[C@H](O)C[C@H]4 [C@@H]3 CCc2c([2 H])c1O
IUPAC navn (8 R,9 S,13 S,14 S,16 R,17 R)-2,4-dideuterio-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
Molekylvægt (g/mol) 290.39
Formel vægt 290.1851 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity