Få mere at vide
(16alpha)-21-Chloro-17-[(2-furanylcarbonyl)oxy]-16-methyl-pregna-1,4-diene-3,11,20-trione Mometasone, TRC
Artikelnummer. 17961365
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17961365 2.5 mg 2.5mg
17953585 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17961365 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3665202.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1305334-31-9
Molekylær formel C27 H29 Cl O6
Molekylvægt (g/mol) 484.97
Synonym (16alpha)-21-Chloro-17-[(2-furanylcarbonyl)oxy]-16-methylpregna-1,4-diene-3,11,20-trione, Mometasone Furoate Imp. C (EP), 21-Chloro-16alpha-methyl-3,11,20-trioxopregna-1,4-dien-17-yl Furan-2-carboxylate
IUPAC navn [(8S,9S,10R,13S,14S,16R,17R)-17-(2-chloroacetyl)-10,13,16-trimethyl-3,11-dioxo-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl
Tekniske data
Kemisk navn eller materiale 21-Chloro-16alpha-methyl-3,11,20-trioxopregna-1,4-dien-17-yl Furan-2-carboxylate
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C27H29ClO6/c1-15-11-19-18-7-6-16-12-17(29)8-9-25(16,2)23(18)20(30)13-26(19,3)27(15,22(31)14-28)34-24(32)21-5-4-10-33-21/h4-5,8-10,12,15,18-19,23H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,23-,25+,26+,27+/m1/s1
Formel vægt 484.1653
Procent renhed >95%