Få mere at vide
16-Methanethiosulfonyl Hexadecanoic Acid, TRC
Artikelnummer. 30480790
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480790 10 mg
30466000 5 mg
30473023 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480790 Leverandør Toronto Research Chemicals Leverandørnr. TRCM25994010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 887406-81-7
Molekylær formel C17 H34 O4 S2
Molekylvægt (g/mol) 366.58
IUPAC navn 16-methylsulfanylsulfonylhexadecanoic acid
SMIL CSS(=O)(=O)CCCCCCCCCCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale 16-Methanethiosulfonyl Hexadecanoic Acid
Mængde 10 mg
InChI formel InChI=1S/C17H34O4S2/c1-22-23(20,21)16-14-12-10-8-6-4-2-3-5-7-9-11-13-15-17(18)19/h2-16H2,1H3,(H,18,19)
Formel vægt 366.1899