Få mere at vide
16-Epiestriol, Ultra Pure (contains less than 0.3% Estriol), TRC
Artikelnummer. 30376099
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30376099 2.5 mg 2.5mg
30372287 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30376099 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5865012.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 547-81-9
Molekylær formel C18 H24 O3
Molekylvægt (g/mol) 288.38
Synonym Estra-1,3,5(10)-triene-3,16beta,17beta-triol, Estriol Imp. F (EP), 16-epi-Estriol, Estra-1,3,5(10)-triene-3,16,17-triol, (16β,17β)-, Estra-1,3,5(10)-triene-3,16β,17-triol (7CI), Estra-1,3,5(10)-triene-3,16β,17β-triol (8CI), (16β,17β)-Estra-1,3,5(10)-triene-3,16,17-triol, 16-Epiestratriol, 16-Epiestriol, 16β,17β-Estriol, 16β-Hydroxyestradiol, Actriol, Epiestriol, Epioestriol, NSC 26646
IUPAC navn (8R,9S,13S,14S,16S,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
SMIL C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@H](O)[C@@H]2O
Tekniske data
Kemisk navn eller materiale 16-Epiestriol
Mængde 2.5 mg
InChI formel InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16+,17+,18+/m1/s1
Formel vægt 288.1725