Få mere at vide
16-Epi-deacetyl-fusidic Acid, TRC
Artikelnummer. 30011572
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011572 250 mg
30009517 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011572 Leverandør Toronto Research Chemicals Leverandørnr. TRCE583000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5951-83-7
Molekylær formel C29 H46 O5
Molekylvægt (g/mol) 474.6725
Synonym 29-Nordammara-17(20),24-dien-21-oic acid, 3,11,16-trihydroxy-, (3α,4α,8α,9β,11α,13α,14β,16α,17Z)-, 16-Epifusidic acid, deacetyl- (7CI), 29-Nor-8α,9β,13α,14β-dammara-17(20),24-dien-21-oic acid, 3α,11α,16α-trihydroxy-, (Z)- (8CI), (3α,4α,8α,9β,11α,13α,14β,16α,17Z)-3,11,16-Trihydroxy-29-nordammara-17(20),24-dien-21-oic acid, 16-Epideacetylfusidic acid, NSC 319631, ent-(17Z)-3β,11β,16β-Trihydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dien-21-oic acid, 16-epi-Deacetylfusidic acid
IUPAC navn (2Z)-6-methyl-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16R)-3,11,16-trihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]hept-5-enoic acid
SMIL C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](O)C[C@]34C
Tekniske data
Kemisk navn eller materiale 16-epi-Deacetylfusidic Acid
Mængde 250 mg
InChI formel InChI=1S/C29H46O5/c1-16(2)8-7-9-18(26(33)34)24-20-14-22(31)25-27(4)12-11-21(30)17(3)19(27)10-13-28(25,5)29(20,6)15-23(24)32/h8,17,19-23,25,30-32H,7,9-15H2,1-6H3,(H,33,34)/b24-18-/t17-,19-,20-,21+,22+,23+,25-,27-,28-,29-/m0/s1
Formel vægt 474.3345