Få mere at vide
16-Dehydroprogesterone, Mikromol™
Artikelnummer. 30049930
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049930 25 mg
30050666 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049930 Leverandør Mikromol Leverandørnr. MM0476.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
16-Dehydroprogesterone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 16-Dehydroprogesterone
CAS 1096-38-4
Analyt- eller komponentnavne 16-Dehydroprogesterone
Forsendelsestilstand Room Temperature
Molekylær formel C21 H28 O2
Certificeringer/compliance ISO 17025
Synonym Pregna-4,16-diene-3,20-dione, 16,17-Didehydroprogesterone, 16-Dehydroprogesterone, 3,20-Dioxopregna-4,16-diene, NSC 11037, NSC 9785, Δ16-Progesterone, Δ4,16-Pregnadiene-3,20-dione
InChI formel InChI=1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h6,12,16,18-19H,4-5,7-11H2,1-3H3/t16-,18-,19-,20-,21+/m0/s1
SMIL CC(=O)C1=CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn (8R,9S,10R,13S,14S)-17-acetyl-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 312.45
Mængde 25 mg
Formel vægt 312.2089 g/mol