Få mere at vide
16-Dehydropregnolone, TRC
Artikelnummer. 30363966
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363966 1 g
30351318 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363966 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2330351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1162-53-4
Molekylær formel C21 H30 O2
Molekylvægt (g/mol) 314.46
Synonym (3β)-3-Hydroxy-pregna-5,16-dien-20-one, Pregna-5,16-dien-20-one, 3β-hydroxy- (6CI,8CI), (-)-Pregnadienolone, (3β)-3-Hydroxypregna-5,16-dien-20-one, 16-Dehydropregnenolone, 16-Dehydropregnolone, 3β-Hydroxy-20-oxopregna-5,16-diene, 3β-Hydroxy-δ5,16-pregnadien-20-one, 3β-Hydroxypregna-5,16-dien-20-one, NSC 15467, Pregna-5,16-dien-3β-ol-20-one, Pregnadienolone, δ16-Pregnenolone
IUPAC navn 1-[(3S,8R,9S,10R,13S,14S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMIL CC(=O)C1=CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale 16-Dehydropregnolone
Mængde 1 g
InChI formel InChI=1S/C21H30O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,6,15-16,18-19,23H,5,7-12H2,1-3H3/t15-,16-,18-,19-,20-,21+/m0/s1
Formel vægt 314.2246