Få mere at vide
16-Dehydropregnenolone Acetate Oxime, Mikromol™
Artikelnummer. 30051219
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051219 25 mg
17951596 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051219 Leverandør Mikromol Leverandørnr. MM0651.020025
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
16-Dehydropregnenolone Acetate Oxime
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 16-Dehydropregnenolone Acetate Oxime
CAS 2174-13-2
Analyt- eller komponentnavne 16-Dehydropregnenolone Acetate Oxime
Forsendelsestilstand Room Temperature
Molekylær formel C23 H33 N O3
Certificeringer/compliance ISO 9001
Synonym 16-Dehydropregnenolone Acetate Oxime
InChI formel InChI=1S/C23H33NO3/c1-14(24-26)19-7-8-20-18-6-5-16-13-17(27-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,7,17-18,20-21,26H,6,8-13H2,1-4H3/b24-14-/t17-,18-,20-,21-,22-,23+/m0/s1
SMIL CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4\C(=N/O)\C)[C@@H]3CC=C2C1
IUPAC navn [(3S,8R,9S,10R,13S,14S)-17-[(Z)-N-hydroxy-C-methylcarbonimidoyl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molekylvægt (g/mol) 371.51
Mængde 25 mg
Formel vægt 371.246 g/mol