Learn More
Ammonium chloride, BAKER ANALYZED™ ACS, J.T. Baker™
Click to view available options
Quantity:
500 g
1 kg
Specifications
Melting Point | 335°C |
pH | 4.5 to 5.5 (5% Solution at 25°C) |
Quantity | 500 g |
Assay Percent Range | min. 99.5% |
IUPAC Name | amine hydrochloride |
Packaging | Plastic bottle for solids |
Residue after Ignition | max. 0.01% |
Density | 1.5256 g/cm³ (20°C) |