Få mere at vide
(14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine Maleate, Mikromol™
Artikelnummer. 30052722
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30052722 25 mg
1 options
Artikelnummer. 30052722 Leverandør Mikromol Leverandørnr. MM1005.090025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine Maleate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (14bRS)-1,2,3,4,10,14b-Hexahydropyrazino(2,1-a)pyrido(2,3-c)(2)benzazepine Maleate
CAS 135928-31-3
Analyt- eller komponentnavne (14bRS)-1,2,3,4,10,14b-Hexahydropyrazino(2,1-a)pyrido(2,3-c)(2)benzazepine Maleate
Forsendelsestilstand Room Temperature
Molekylær formel C16 H17 N3 . C4 H4 O4
Certificeringer/compliance ISO 17025
Synonym Mirtazapine Imp. D (EP), Mirtazapine USP Related Compound A, Mirtazapine USP RC A, (14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine Maleate, Mirtazapine Imp. D (EP) as Maleate, Mirtazapine Related Compound A as Maleate, Mirtazapine Impurity D as Maleate
InChI formel InChI=1S/C16H17N3.C4H4O4/c1-2-6-14-12(4-1)10-13-5-3-7-18-16(13)19-9-8-17-11-15(14)19;5-3(6)1-2-4(7)8/h1-7,15,17H,8-11H2;1-2H,(H,5,6)(H,7,8)/b;2-1-
SMIL OC(=O)\C=C/C(=O)O.C1CN2C(CN1)c3ccccc3Cc4cccnc24
IUPAC navn (14bRS)-1,2,3,4,10,14b-Hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine Maleate
Molekylvægt (g/mol) 367.4
Mængde 25 mg
Formel vægt 367.1532 g/mol