missing translation for 'onlineSavingsMsg'
Få mere at vide

(14bRS)-1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride (Desmethylmianserin Hydrochloride), Mikromol™

Product Code. 30051020
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30051020 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051020 Supplier Mikromol Supplier No. MM0147.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(14bRS)-1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride (Desmethylmianserin Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Normianserin Hydrochloride
CAS 76134-77-5
Analyt- eller komponentnavne Normianserin Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H18 N2 . Cl H
Certificeringer/compliance ISO 17025
Synonym Mianserin Hydrochloride Imp. E (EP), Mianserin Imp. E (EP), (14bRS)-1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine Hydrochloride, Mianserin Hydrochloride Imp. E (EP) as Hydrochloride, Desmethylmianserin Hydrochloride, Normianserin Hydrochloride, Mianserin Hydrochloride Impurity E as Hydrochloride, Mianserin Impurity E as Hydrochloride
InChI formel InChI=1S/C17H18N2.ClH/c1-3-7-15-13(5-1)11-14-6-2-4-8-16(14)19-10-9-18-12-17(15)19;/h1-8,17-18H,9-12H2;1H
SMIL Cl.C1CN2C(CN1)c3ccccc3Cc4ccccc24
IUPAC navn (14bRS)-1,2,3,4,10,14b-Hexahydrodibenzo[c,f]pyrazino[1,2-a]azepine hydrochloride
Molekylvægt (g/mol) 286.8
Mængde 50 mg
Formel vægt 286.1237 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.