Få mere at vide
13Beta-Ethyl-11-methylenegon-4-en-17-one, TRC
Artikelnummer. 30370175
Skift visning
Mængde:
10 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370175 500 mg
30369607 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30370175 Leverandør Toronto Research Chemicals Leverandørnr. TRCE925095500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54024-21-4
Molekylær formel C20 H28 O
Molekylvægt (g/mol) 284.44
Synonym 13-Ethyl-11-methylene-gon-4-en-17-one, (+)-13β-Ethyl-11-methylenegon-4-en-17-one, 13β-Ethyl-11-methylenegon-4-en-17-one, Desogestrel Imp. C (EP)
IUPAC navn (8S,9S,10R,13S,14S)-13-ethyl-11-methylidene-1,2,3,6,7,8,9,10,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-one
SMIL CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CCC2=O
Tekniske data
Kemisk navn eller materiale 13-beta-Ethyl-11-methylenegon-4-en-17-one
Mængde 500 mg
InChI formel InChI=1S/C20H28O/c1-3-20-12-13(2)19-15-7-5-4-6-14(15)8-9-16(19)17(20)10-11-18(20)21/h6,15-17,19H,2-5,7-12H2,1H3/t15-,16-,17-,19+,20-/m0/s1
Formel vægt 284.214