Få mere at vide
13-O-(Triethylsilyl) Baccatin III, TRC
Artikelnummer. 30455934
Skift visning
Mængde:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455934 25 mg
30450429 250 mg
30449993 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455934 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77793025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 208406-86-4
Molekylær formel C37 H52 O11 Si
Molekylvægt (g/mol) 700.89
Synonym 7,11-Methano-5H-cyclodeca[3,4]benz[1,2-b]oxet-5-one, 6,12b-bis(acetyloxy)-12-(benzoyloxy)-1,2a,3,4,4a,6,9,10,11,12,12a,12b-dodecahydro-4,11-dihydroxy-4a,8,13,13-tetramethyl-9-[(triethylsilyl)oxy]-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-, 13-O-(Triethylsilyl)baccatin III, 13-Tes-baccatin III
IUPAC navn [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4,12-diacetyloxy-1,9-dihydroxy-10,14,17,17-tetramethyl-11-oxo-15-triethylsilyloxy-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
SMIL CC[Si](CC)(CC)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@@H]4[C@@]5(CO[C@@H]5C[C@H](O)[C@@]4(C)C(=O)[C@H](OC(=O)C)C(=C1C)C2(C)C)OC(=O)C
Tekniske data
Kemisk navn eller materiale 13-O-(Triethylsilyl) Baccatin III
Mængde 25 mg
InChI formel InChI=1S/C37H52O11Si/c1-10-49(11-2,12-3)48-25-19-37(43)32(46-33(42)24-16-14-13-15-17-24)30-35(9,26(40)18-27-36(30,20-44-27)47-23(6)39)31(41)29(45-22(5)38)28(21(25)4)34(37,7)8/h13-17,25-27,29-30,32,40,43H,10-12,18-20H2,1-9H3/t25-,26-,27+,29+,30-,32-,35+,36-,37+/m0/s1
Formel vægt 700.3279