missing translation for 'onlineSavingsMsg'
Få mere at vide

13-cis-Retinyl Acetate (85%), TRC

Artikelnummer. 30421629
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421629 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30421629 Leverandør Toronto Research Chemicals Leverandørnr. TRCR23990525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 13-cis-Retinyl Acetate
CAS 34356-31-5
Molekylær formel C22 H32 O2
Mængde 25 mg
Synonym Retinol','15-acetate','13-cis-, Retinol','acetate','13-cis- (8CI,9CI), 13-cis-Retinol acetate, 13-cis-Retinyl acetate, 13-cis-Vitamin A acetate
InChI formel InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14-
SMIL CC(=O)OC\C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C
IUPAC navn [(2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
Molekylvægt (g/mol) 328.49
Formel vægt 328.2402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.