Få mere at vide
13-cis-Retinoic Acid-d5 Ethyl Ester, TRC
Artikelnummer. 30422607
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422607 1 mg
30424444 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422607 Leverandør Toronto Research Chemicals Leverandørnr. TRCR2502971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 13-cis-Retinoic Acid-d5 Ethyl Ester
CAS 78995-96-7
Molekylær formel C22 H27 D5 O2
Mængde 1 mg
Synonym (8Z)-3,7-Dimethyl-9-(2,6,6-trimethyl-cyclohex-1-enyl)-nona-2,4,6,8-tetraenoic Acid-d5 Ethyl Ester, (13Z)-Ethyl Retinoate-d5
InChI formel InChI=1S/C22H32O2/c1-7-24-21(23)16-18(3)11-8-10-17(2)13-14-20-19(4)12-9-15-22(20,5)6/h8,10-11,13-14,16H,7,9,12,15H2,1-6H3/b11-8+,14-13+,17-10+,18-16-/i4D3,12D2
SMIL [2H]C([2H])([2H])C1=C(\C=C\C(=C\C=C\C(=C/C(=O)OCC)\C)\C)C(C)(C)CCC1([2H])[2H]
IUPAC navn ethyl (2Z,4E,6E,8E)-9-[3,3-dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]-3,7-dimethyl-nona-2,4,6,8-tetraenoate
Molekylvægt (g/mol) 333.51909
Formel vægt 333.2716