Få mere at vide
12-Deoxy Roxithromycin, TRC
Artikelnummer. 30368501
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368501 1 mg
30368394 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30368501 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2540001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 425365-65-7
Molekylær formel C41H76N2O14
Molekylvægt (g/mol) 821.05
Synonym (9E)-12-Deoxy-9-[O-[(2-methoxyethoxy)methyl]oxime] Erythromycin
SMIL O=C([C@H](C)[C@@H](O[C@@H]1O[C@@H](C)[C@H](O)[C@](C)(OC)C1)[C@H](C)[C@@H](O[C@H]2[C@H](O)[C@@H](N(C)C)C[C@@H](C)O2)[C@@](O)(C)C[C@H]/3C)O[C@H](CC)[C@H](C)[C@H](O)[C@@H](C)C3=N\OCOCCOC
Tekniske data
Kemisk navn eller materiale 12-Deoxy Roxithromycin
Mængde 1 mg
InChI formel InChI=1S/C41H76N2O14/c1-15-30-24(4)33(44)25(5)32(42-52-21-51-17-16-49-13)22(2)19-40(9,48)37(57-39-34(45)29(43(11)12)18-23(3)53-39)26(6)35(27(7)38(47)55-30)56-31-20-41(10,50-14)36(46)28(8)54-31/h22-31,33-37,39,44-46,48H,15-21H2,1-14H3/b42-32+/t22-,23?,24+,25+,26+,27-,28?,29?,30-,31?,33+,34?,35+,36?,37-,39?,40-,41?/m1/s1