missing translation for 'onlineSavingsMsg'
Få mere at vide

12-Aminododecyl Methanethiosulfonate Hydrochloride, TRC

Artikelnummer. 30320085
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320085 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30320085 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6088141MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 12-Aminododecyl Methanethiosulfonate Hydrochloride
CAS 1246816-18-1
Molekylær formel C13H30ClNO2S2
Mængde 1 mg
InChI formel InChI=1S/C13H29NO2S2.ClH/c1-18(15,16)17-13-11-9-7-5-3-2-4-6-8-10-12-14;/h2-14H2,1H3;1H
SMIL Cl.CS(=O)(=O)SCCCCCCCCCCCCN
IUPAC navn 12-methylsulfonylsulfanyldodecan-1-amine;hydrochloride
Molekylvægt (g/mol) 331.97
Formel vægt 331.1406
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.