Få mere at vide
11Beta-Hydroxy Progesterone, TRC
Artikelnummer. 30138154
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138154 500 mg
30132875 250 mg
30008292 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138154 Leverandør Toronto Research Chemicals Leverandørnr. TRCB249680500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 600-57-7
Molekylær formel C21 H30 O3
Synonym 11β-Hydroxy-pregn-4-ene-3,20-dione∣21-Deoxycorticosterone
Tekniske data
Kemisk navn eller materiale 11beta-Hydroxyprogesterone
Mængde 500 mg
Forsendelsestilstand Room Temperature
Formel vægt 330.2195
Renhed >95% (HPLC)
Molekylvægt (g/mol) 330.46