Få mere at vide
11beta-Hydroxy-6alpha-methyl-1,4-androstadiene-3,17-dione, TRC
Artikelnummer. 30012629
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012629 50 mg
30010043 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30012629 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94643550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61919-52-6
Molekylær formel C20 H26 O3
Molekylvægt (g/mol) 314.42
Synonym Androsta-1,4-diene-3,17-dione, 11-hydroxy-6-methyl-, (6alpha,11beta)- (9CI), Androsta-1,4-diene-3,17-dione, 11beta-hydroxy-6alpha-methyl- (7CI), 11beta-Hydroxy-6alpha-methyl-1,4-androstadiene-3,17-dione, 11beta-Hydroxy-6alpha-methylandrosta-1,4-diene-3,17-dione
IUPAC navn (6S,8S,9S,10R,11S,13S,14S)-11-hydroxy-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-dione
SMIL C[C@H]1C[C@H]2[C@@H]3CCC(=O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
Tekniske data
Kemisk navn eller materiale 11beta-Hydroxy-6alpha-methylandrosta-1,4-diene-3,17-dione
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C20H26O3/c1-11-8-13-14-4-5-17(23)20(14,3)10-16(22)18(13)19(2)7-6-12(21)9-15(11)19/h6-7,9,11,13-14,16,18,22H,4-5,8,10H2,1-3H3/t11-,13-,14-,16-,18+,19-,20-/m0/s1
Formel vægt 314.1882
Procent renhed >95%