Få mere at vide
(11Beta)-9-Fluoro-11,21-dihydroxy-16-methyl-pregna-1,4,16-triene-3,20-dione, TRC
Artikelnummer. 30132730
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132730 100 mg
17910385 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132730 Leverandør Toronto Research Chemicals Leverandørnr. TRCF591115100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59860-99-0
Molekylær formel C22 H27 F O4
Molekylvægt (g/mol) 374.45
Synonym Pregna-1,4,16-triene-3,20-dione, 9-fluoro-11,21-dihydroxy-16-methyl-, (11β)-, (11β)-9-Fluoro-11,21-dihydroxy-16-methylpregna-1,4,16-triene-3,20-dione, 9-Fluoro-11β,21-dihydroxy-16-methylpregna-1,4,16-triene-3,20-dione, Pregna-1,4,16-triene-3,20-dione, 9-fluoro-11,21-dihydroxy-16-methyl-, (11β)-, (11β)-9-Fluoro-11,21-dihydroxy-16-methylpregna-1,4,16-triene-3,20-dione, 9-Fluoro-11β,21-dihydroxy-16-methylpregna-1,4,16-triene-3,20-dione, 9-Fluoro-11beta,21-dihydroxy-16-methylpregna-1,4,16-triene-3,20-dione
IUPAC navn (8S,9R,10S,11S,13S,14S)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-7,8,11,12,14,15-hexahydro-6H-cyclopenta[a]phenanthren-3-one
SMIL CC1=C(C(=O)CO)[C@@]2(C)C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]2C1
Tekniske data
Kemisk navn eller materiale (11beta)-9-Fluoro-11,21-dihydroxy-16-methylpregna-1,4,16-triene-3,20-dione
Mængde 100 mg
InChI formel InChI=1S/C22H27FO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-21(13,3)22(15,23)18(27)10-20(16,2)19(12)17(26)11-24/h6-7,9,15-16,18,24,27H,4-5,8,10-11H2,1-3H3/t15-,16-,18-,20-,21-,22-/m0/s1
Formel vægt 374.1893