Få mere at vide
(11Beta,16Alpha)-9-Chloro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione (Mometasone Impurity M), TRC
Artikelnummer. 30352624
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30352624 50 mg
30352623 25 mg
30349269 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30352624 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37084050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57780-86-6
Molekylær formel C22H29ClO4
Molekylvægt (g/mol) 392.92
Synonym Pregna-1,4-diene-3,20-dione, 9-chloro-11,17-dihydroxy-16-methyl-, (11β,16α)- (9CI), 9-Chloro-11β,17-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, Mometasone Furoate Imp. M (EP)
IUPAC navn (8S,9R,10S,11S,13S,14S,16R,17R)-17-acetyl-9-chloro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)C
Tekniske data
Kemisk navn eller materiale (11beta,16alpha)-9-Chloro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione (Mometasone Impurity M)
Mængde 50 mg
InChI formel InChI=1S/C22H29ClO4/c1-12-9-17-16-6-5-14-10-15(25)7-8-19(14,3)21(16,23)18(26)11-20(17,4)22(12,27)13(2)24/h7-8,10,12,16-18,26-27H,5-6,9,11H2,1-4H3/t12-,16+,17+,18+,19+,20+,21+,22+/m1/s1
Formel vægt 392.1754